Merck
  • Prognostic value of volumetric FDG PET/CT parameters in patients with oral tongue squamous cell carcinoma who were treated by superselective intra-arterial chemoradiotherapy.
Prognostic value of volumetric FDG PET/CT parameters in patients with oral tongue squamous cell carcinoma who were treated by superselective intra-arterial chemoradiotherapy.
Japanese journal of radiology (2017-10-19)
Satoko Suzuki-Shibata, Yayoi Yamamoto, Tetsuo Yoshida, Nobutaka Mizoguchi, Tetsuo Nonaka, Akira Kubota, Hiroto Narimatsu, Yohei Miyagi, Toshiaki Kobayashi, Tomohiro Kaneta, Tomio Inoue
ZUSAMMENFASSUNG
This study aimed to evaluate the predictive and prognostic value of FDG PET/CT-based volumetric parameters in patients with oral tongue squamous cell carcinoma (OTSCC) treated by superselective intra-arterial chemoradiotherapy (IA-CRT). We conducted a retrospective study including 33 patients with biopsy-proven OTSCC between May 2007 and February 2016. All of the patients were treated by IA-CRT. Pretreatment SUV The median follow-up for surviving patients was 40.7 months (range 6.0-107.5 months). In univariate and multivariate analyses, SUV FDG PET/CT-based volumetric parameters may be significant prognostic markers for survival of patients with OTSCC who are treated by IA-CRT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluorescein di(β-D-galactopyranosid)