Merck
  • A study on amphiphilic fluorinated block copolymer in graphite exfoliation using supercritical CO
A study on amphiphilic fluorinated block copolymer in graphite exfoliation using supercritical CO
Journal of colloid and interface science (2017-09-25)
Young Hyun Kim, Hyang Moo Lee, Sung Wook Choi, In Woo Cheong
ZUSAMMENFASSUNG
In this study, poly(2,2,2-trifluoroethyl methacrylate)-block-poly(4-vinylpyridine) (PTFEMA-b-PVP) was synthesized by stepwise reversible addition-fragmentation chain transfer (RAFT) polymerization for the preparation of graphene by the exfoliation of graphite nanoplatelets (GPs) in supercritical CO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Vinylpyridin, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
4-Cyan-4-(thiobenzoylthio)-pentansäure
Sigma-Aldrich
2,2,2-Trifluorethylmethacrylat, contains 50-200 ppm MEHQ as inhibitor, 99%