- Goats without Prion Protein Display Enhanced Proinflammatory Pulmonary Signaling and Extracellular Matrix Remodeling upon Systemic Lipopolysaccharide Challenge.
Goats without Prion Protein Display Enhanced Proinflammatory Pulmonary Signaling and Extracellular Matrix Remodeling upon Systemic Lipopolysaccharide Challenge.
Frontiers in immunology (2017-12-23)
Øyvind Salvesen, Malin R Reiten, Jorke H Kamstra, Maren K Bakkebø, Arild Espenes, Michael A Tranulis, Cecilie Ersdal
PMID29270176
ZUSAMMENFASSUNG
A naturally occurring mutation in the
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture