Merck
  • Reply to The small molecule CLP257 does not modify activity of the K+-Cl- co-transporter KCC2 but does potentiate GABAA receptor activity.
Reply to The small molecule CLP257 does not modify activity of the K+-Cl- co-transporter KCC2 but does potentiate GABAA receptor activity.
Nature medicine (2017-12-08)
Martin Gagnon, Marc J Bergeron, Jimena Perez-Sanchez, Isabel Plasencia-Fernández, Louis-Etienne Lorenzo, Antoine G Godin, Annie Castonguay, Robert P Bonin, Yves De Koninck
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-K+/Cl- Cotransporter (KCC2)-Antikörper, Upstate®, from rabbit