Merck
  • Palmitoylethanolamide Supplementation during Sensitization Prevents Airway Allergic Symptoms in the Mouse.
Palmitoylethanolamide Supplementation during Sensitization Prevents Airway Allergic Symptoms in the Mouse.
Frontiers in pharmacology (2018-01-10)
Fiorentina Roviezzo, Antonietta Rossi, Elisabetta Caiazzo, Pierangelo Orlando, Maria A Riemma, Valentina M Iacono, Andrea Guarino, Armando Ialenti, Carla Cicala, Alessio Peritore, Raffaele Capasso, Vincenzo Di Marzo, Angelo A Izzo
ZUSAMMENFASSUNG
One important risk factor for the development of asthma is allergen sensitization. Recent increasing evidence suggests a prominent role of mast cells in asthma pathophysiology. Since Palmitoylethanolamide (PEA), an endogenous lipid mediator chemically related to - and co-released with- the endocannabinoid anandamide, behaves as a local autacoid down-regulator of mast cell activation and inflammation, we explored the possible contribution of PEA in allergic sensitization, by using ovalbumin (OVA) as sensitizing agent in the mouse. PEA levels were dramatically reduced in the bronchi of OVA-treated animals. This effect was coupled to a significant up-regulation of CB
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Palmitoylethanolamide