Merck
  • Untargeted metabolomics reveals specific withanolides and fatty acyl glycoside as tentative metabolites to differentiate organic and conventional Physalis peruviana fruits.
Untargeted metabolomics reveals specific withanolides and fatty acyl glycoside as tentative metabolites to differentiate organic and conventional Physalis peruviana fruits.
Food chemistry (2017-11-10)
Sandra M Llano, Ana M Muñoz-Jiménez, Claudio Jiménez-Cartagena, Julián Londoño-Londoño, Sonia Medina
ZUSAMMENFASSUNG
The agronomic production systems may affect the levels of food metabolites. Metabolomics approaches have been applied as useful tool for the characterization of fruit metabolome. In this study, metabolomics techniques were used to assess the differences in phytochemical composition between goldenberry samples produced by organic and conventional systems. To verify that the organic samples were free of pesticides, individual pesticides were analyzed. Principal component analysis showed a clear separation of goldenberry samples from two different farming systems. Via targeted metabolomics assays, whereby carotenoids and ascorbic acid were analyzed, not statistical differences between both crops were found. Conversely, untargeted metabolomics allowed us to identify two withanolides and one fatty acyl glycoside as tentative metabolites to differentiate goldenberry fruits, recording organic fruits higher amounts of these compounds than conventional samples. Hence, untargeted metabolomics technology could be suitable to research differences on phytochemicals under different agricultural management practices and to authenticate organic products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Azoxystrobin, PESTANAL®, analytical standard
Supelco
Zeaxanthin, analytical standard
Supelco
Trifloxystrobin, PESTANAL®, analytical standard
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Supelco
Cymoxanil, PESTANAL®, analytical standard
Supelco
Methomyl, PESTANAL®, analytical standard
Supelco
Cartap -hydrochlorid, PESTANAL®, analytical standard
Supelco
Myclobutanil, PESTANAL®, analytical standard
Supelco
Ethoprophos, PESTANAL®, analytical standard
Supelco
Fenhexamid, PESTANAL®, analytical standard
Supelco
Spiromesifen, PESTANAL®, analytical standard