- N-linked glycosylation of a subunit isoforms is critical for vertebrate vacuolar H
N-linked glycosylation of a subunit isoforms is critical for vertebrate vacuolar H
Journal of cellular biochemistry (2017-07-01)
Sally Esmail, Norbert Kartner, Yeqi Yao, Joo Wan Kim, Reinhart A F Reithmeier, Morris F Manolson
PMID28661051
ZUSAMMENFASSUNG
The a subunit of the V
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-VMA21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution