- Discovery of High-Affinity PDGF-VEGFR Interactions: Redefining RTK Dynamics.
Discovery of High-Affinity PDGF-VEGFR Interactions: Redefining RTK Dynamics.
Scientific reports (2017-11-29)
Spencer B Mamer, Si Chen, Jared C Weddell, Alexandra Palasz, Ashley Wittenkeller, Manu Kumar, P I Imoukhuede
PMID29180757
ZUSAMMENFASSUNG
Nearly all studies of angiogenesis have focused on uni-family ligand-receptor binding, e.g., VEGFs bind to VEGF receptors, PDGFs bind to PDGF receptors, etc. The discovery of VEGF-PDGFRs binding challenges this paradigm and calls for investigation of other ligand-receptor binding possibilities. We utilized surface plasmon resonance to identify and measure PDGF-to-VEGFR binding rates, establishing cut-offs for binding and non-binding interactions. We quantified the kinetics of the recent VEGF-A:PDGFRβ interaction for the first time with K