Merck
  • Serum of patients with antiphospholipid syndrome induces adhesion molecules in endothelial cells.
Serum of patients with antiphospholipid syndrome induces adhesion molecules in endothelial cells.
Atherosclerosis. Supplements (2017-11-04)
Bettina Engel, Gregor Müller, Beate Roch, Hans-Egbert Schröder, Martin Aringer, Stefan R Bornstein, Henning Morawietz
ZUSAMMENFASSUNG
The antiphospholipid syndrome (APS) is a systemic auto-immune disease with an unclear pathophysiology. The aim of our study was to understand the development of APS on a cellular level. Therefore, we analyzed the influence of human serum of APS patients on endothelial expression of specific genes and proteins in comparison to a control group. In this study, we analyzed the expression of ICAM-1, VCAM-1, E-selectin and annexin V in primary cultures of human umbilical vein endothelial cells (HUVEC) in response to 10% (v/v) serum of control patients (n = 6), patients with systemic lupus erythematosus (SLE) and no APS (n = 4) or APS patients (n = 9) for 24 h. Total RNA was prepared from confluent endothelial cell layers and mRNA expression of ICAM-1, VCAM-1 and E-selectin was analyzed by reverse transcription polymerase-chain reaction (RT-PCR). The protein expression was determined by Western blot. Serum protein concentrations of soluble forms of adhesion molecules sICAM-1 and sVCAM-1 were quantified by ELISA. Gene expression data were correlated with clinical parameters. The mRNA expression of ICAM-1 was increased in cells incubated with serum from APS patients (166 ± 22% of control; P = 0.023). Serum of patients with (SLE)/no APS caused a 1.4-fold higher ICAM-1 mRNA level. Western blot analysis showed an increase in protein expression of adhesion molecules ICAM-1 (260 ± 49%; P = 0.011) and VCAM-1 (357 ± 97%; P = 0.023) in cells that were incubated with serum from APS patients. Plasma analysis showed elevated levels of sVCAM-1 in APS patients (189 ± 34%; P = 0.045) compared to the levels measured in the control group. The sVCAM-1 plasma level was correlating with the frequency of abortions. An augmented expression of endothelial adhesion molecules is involved in the pathophysiology of patients with antiphospholipid syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O