Merck
  • Unspliced XBP1 Confers VSMC Homeostasis and Prevents Aortic Aneurysm Formation via FoxO4 Interaction.
Unspliced XBP1 Confers VSMC Homeostasis and Prevents Aortic Aneurysm Formation via FoxO4 Interaction.
Circulation research (2017-11-02)
Guizhen Zhao, Yi Fu, Zeyu Cai, Fang Yu, Ze Gong, Rongbo Dai, Yanhua Hu, Lingfang Zeng, Qingbo Xu, Wei Kong
ZUSAMMENFASSUNG
Although not fully understood, the phenotypic transition of vascular smooth muscle cells exhibits at the early onset of the pathology of aortic aneurysms. Exploring the key regulators that are responsible for maintaining the contractile phenotype of vascular smooth muscle cells (VSMCs) may confer vascular homeostasis and prevent aneurysmal disease. XBP1 (X-box binding protein 1), which exists in a transcriptionally inactive unspliced form (XBP1u) and a spliced active form (XBP1s), is a key component in response to endoplasmic reticular stress. Compared with XBP1s, little is known about the role of XBP1u in vascular homeostasis and disease. We aim to investigate the role of XBP1u in VSMC phenotypic switching and the pathogenesis of aortic aneurysms. XBP1u, but not XBP1s, was markedly repressed in the aorta during the early onset of aortic aneurysm in both angiotensin II-infused apolipoprotein E knockout (ApoE Our study revealed the pivotal role of the XBP1u-FoxO4-myocardin axis in maintaining the VSMC contractile phenotype and providing protection from aortic aneurysm formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology