Merck
The parasite
The Journal of cell biology (2017-10-27)
Julia D Romano, Sabrina J Nolan, Corey Porter, Karen Ehrenman, Eric J Hartman, Ru-Ching Hsia, Isabelle Coppens
ZUSAMMENFASSUNG
Many intracellular pathogens subvert host membrane trafficking pathways to promote their replication.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%