Merck
Synthetic Organic Electrochemical Methods Since 2000: On the Verge of a Renaissance.
Chemical reviews (2017-10-11)
Ming Yan, Yu Kawamata, Phil S Baran
ZUSAMMENFASSUNG
Electrochemistry represents one of the most intimate ways of interacting with molecules. This review discusses advances in synthetic organic electrochemistry since 2000. Enabling methods and synthetic applications are analyzed alongside innate advantages as well as future challenges of electroorganic chemistry.