Merck
Activity-dependent expression of Channelrhodopsin at neuronal synapses.
Nature communications (2017-11-22)
Francesco Gobbo, Laura Marchetti, Ajesh Jacob, Bruno Pinto, Noemi Binini, Federico Pecoraro Bisogni, Claudia Alia, Stefano Luin, Matteo Caleo, Tommaso Fellin, Laura Cancedda, Antonino Cattaneo
ZUSAMMENFASSUNG
Increasing evidence points to the importance of dendritic spines in the formation and allocation of memories, and alterations of spine number and physiology are associated to memory and cognitive disorders. Modifications of the activity of subsets of synapses are believed to be crucial for memory establishment. However, the development of a method to directly test this hypothesis, by selectively controlling the activity of potentiated spines, is currently lagging. Here we introduce a hybrid RNA/protein approach to regulate the expression of a light-sensitive membrane channel at activated synapses, enabling selective tagging of potentiated spines following the encoding of a novel context in the hippocampus. This approach can be used to map potentiated synapses in the brain and will make it possible to re-activate the neuron only at previously activated synapses, extending current neuron-tagging technologies in the investigation of memory processes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Anti-GluR1-NT(-NT)-Antikörper, Klon RH95, clone RH95, from mouse
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution