Merck
  • Copper (II) complexes of bidentate ligands exhibit potent anti-cancer activity regardless of platinum sensitivity status.
Copper (II) complexes of bidentate ligands exhibit potent anti-cancer activity regardless of platinum sensitivity status.
Investigational new drugs (2017-07-25)
Mohamed Wehbe, Cody Lo, Ada W Y Leung, Wieslawa H Dragowska, Gemma M Ryan, Marcel B Bally
ZUSAMMENFASSUNG
Insensitivity to platinum, either through inherent or acquired resistance, is a major clinical problem in the treatment of many solid tumors. Here, we explored the therapeutic potential of diethyldithiocarbamate (DDC), pyrithione (Pyr), plumbagin (Plum), 8-hydroxyquinoline (8-HQ), clioquinol (CQ) copper complexes in a panel of cancer cell lines that differ in their sensitivity to platins (cisplatin/carboplatin) using a high-content imaging system. Our data suggest that the copper complexes were effective against both platinum sensitive (IC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Plumbagin aus Plumbago indica
Sigma-Aldrich
Diethyl-1,4-dihydro-2,4,6-trimethyl-3,5-pyridindicarboxylat, 99%
Sigma-Aldrich
2-Mercaptopyridin-N-oxid, 99%