Merck
  • E-cadherin cleavage by MT2-MMP regulates apical junctional signaling and epithelial homeostasis in the intestine.
E-cadherin cleavage by MT2-MMP regulates apical junctional signaling and epithelial homeostasis in the intestine.
Journal of cell science (2017-10-25)
Jesús Gómez-Escudero, Vanessa Moreno, Mara Martín-Alonso, M Victoria Hernández-Riquer, Tamar Feinberg, Ángel Colmenar, Enrique Calvo, Emilio Camafeita, Fernando Martínez, Menno J Oudhoff, Stephen J Weiss, Alicia G Arroyo
ZUSAMMENFASSUNG
Cadherin-based intercellular adhesions are essential players in epithelial homeostasis, but their dynamic regulation during tissue morphogenesis and remodeling remain largely undefined. Here, we characterize an unexpected role for the membrane-anchored metalloproteinase MT2-MMP in regulating epithelial cell quiescence. Following co-immunoprecipitation and mass spectrometry, the MT2-MMP cytosolic tail was found to interact with the zonula occludens protein-1 (ZO-1) at the apical junctions of polarized epithelial cells. Functionally, MT2-MMP localizes in the apical domain of epithelial cells where it cleaves E-cadherin and promotes epithelial cell accumulation, a phenotype observed in 2D polarized cells as well as 3D cysts. MT2-MMP-mediated cleavage subsequently disrupts apical E-cadherin-mediated cell quiescence resulting in relaxed apical cortical tension favoring cell extrusion and re-sorting of Src kinase activity to junctional complexes, thereby promoting proliferation. Physiologically, MT2-MMP loss of function alters E-cadherin distribution, leading to impaired 3D organoid formation by mouse colonic epithelial cells ex vivo and reduction of cell proliferation within intestinal crypts in vivo Taken together, these studies identify an MT2-MMP-E-cadherin axis that functions as a novel regulator of epithelial cell homeostasis in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Ezrin Antibody, Upstate®, from rabbit
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP15