Merck
  • Tryptophan Catabolism Restricts IFN-??Expressing Neutrophils and Clostridium difficile Immunopathology
Tryptophan Catabolism Restricts IFN-??Expressing Neutrophils and Clostridium difficile Immunopathology
Journal of Immunology (2014)
Mohamad El-Zaatari