Merck
  • Down-regulation of p16 and MGMT promotes the anti-proliferative and pro-apoptotic effects of 5-Aza-dC and radiation on cervical cancer cells.
Down-regulation of p16 and MGMT promotes the anti-proliferative and pro-apoptotic effects of 5-Aza-dC and radiation on cervical cancer cells.
Cell biochemistry and function (2017-11-17)
Guan-di Chen, De-Ying Qian, Zhi-Gang Li, Ge-Ying Fan, Ke-Li You, Yi-Long Wu
ZUSAMMENFASSUNG
Cervical cancer is one of the most common malignancies of the female reproductive system. Therefore, it is critical to investigate the molecular mechanisms involved in the development and progression of cervical cancer. In this study, we stimulated cervical cancer cells with 5-aza-2'-deoxycytidine (5-Aza-dC) and found that this treatment inhibited cell proliferation and induced apoptosis; additionally, methylation of p16 and O-6-methylguanine-DNA methyltransferase (MGMT) was reversed, although their expression was suppressed. 5-Aza-dC inhibited E6 and E7 expression and up-regulated p53, p21, and Rb expression. Cells transfected with siRNAs targeting p16 and MGMT as well as cells stimulated with 5-Aza-dC were arrested in S phase, and the expression of p53, p21, and Rb was up-regulated more significantly. However, when cells were stimulated with 5-Aza-dC after transfection with siRNAs targeting p16 and MGMT, proliferation decreased significantly, and the percentage of cells in the sub-G1 peak and in S phase was significantly increased, suggesting a marked increase in apoptosis. But E6 and E7 overexpression could rescue the observed effects in proliferation. Furthermore, X-ray radiation caused cells to arrest in G2/M phase, but cells transfected with p16- and MGMT-targeted siRNAs followed by X-ray radiation exhibited a significant decrease in proliferation and were shifted toward the sub-G1 peak, also indicating enhanced apoptosis. In addition, the effects of 5-Aza-dC and X-ray radiation were most pronounced when MGMT expression was down-regulated. Therefore, down-regulation of p16 and MGMT expression enhances the anti-proliferative effects of 5-Aza-dC and X-ray radiation. This discovery may provide novel ideas for the treatment of cervical cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbisulfit -Lösung, purum, ~40%