- Identification of polymorphisms in
Identification of polymorphisms in
Asian-Australasian journal of animal sciences (2017-08-22)
Hasina Sultana, Dongwon Seo, Nu-Ri Choi, Md Shamsul Alam Bhuiyan, Seung Hwan Lee, Kang-Nyeong Heo, Jun-Heon Lee
PMID28823136
ZUSAMMENFASSUNG
The aim of this study was to investigate the effect of single nucleotide polymorphisms (SNPs) of the melanogenesis associated transcription factor ( Five Asian duck varieties (black Korean native, white Korean native, commercial Peking, Nageswari, and Bangladeshi Deshi white ducks) were investigated to examine the polymorphisms associated with plumage colors. Among previously identified SNPs, three synonymous SNPs and one indel of Two synonymous SNPs (c.114T>G and c.147T>C) and a 14-bp indel (GCTGCAAAC AGATG) in intron 7 of The results of this study provide a basis for further investigations of the associations between polymorphisms and plumage color phenotypes in Asian duck breeds.
MATERIALIEN