Merck
  • The Impact of the Progressive Efficiency Test on a Rowing Ergometer on White Blood Cells Distribution and Clinical Chemistry Changes in Paralympic Rowers During the Preparatory Stage Before the Paralympic Games in Rio, 2016 - A Case Report.
The Impact of the Progressive Efficiency Test on a Rowing Ergometer on White Blood Cells Distribution and Clinical Chemistry Changes in Paralympic Rowers During the Preparatory Stage Before the Paralympic Games in Rio, 2016 - A Case Report.
Journal of human kinetics (2018-01-18)
Robert Nowak, Rafał Buryta, Krzysztof Krupecki, Tomasz Zając, Marek Zawartka, Patrizia Proia, Dorota Kostrzewa-Nowak
ZUSAMMENFASSUNG
There is a large gap in knowledge regarding research on post-exercise blood changes in disabled athletes. There are relatively few data on adaptive mechanisms to exercise in disabled athletes, including disabled rowers. Two rowers from a Polish adaptive rowing settle TAMix2x that qualified for the Paralympic Games in Rio, 2016 took part in this study. They performed a progressive test on a rowing ergometer until exhaustion. The cardiorespiratory fitness measures, complete blood count, white blood cells' distribution and 30 clinical chemistry variables describing laboratory diagnostic profiles and general health were determined. The extreme effort induced changes in all studied metabolites (glucose, creatinine, urea, uric acid, total and direct bilirubin), albumin, total protein levels in both participants. Furthermore, a post-exercise increase in aspartate transaminase activity, yet a 2-fold decrease during the recovery time in both rowers were found. White blood cell count increased 2-fold after the test. The percentages of natural killer cells were higher and total T lymphocytes were lower after the exercise protocol. There were higher percentages of suppressor/cytotoxic and lower percentages of helper/inducer T lymphocyte subsets in both studied rowers. No changes in B lymphocytes distribution were observed. Lack of inflammatory symptoms during the experiment suggests a high level of rowers' biological adaptation to the physical effort. The different changes in physiological, biochemical and immunological variables are related to the adaptive mechanism to physical exercise allowing for improvement of performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Biphenyl-4,4′-dithiol, 95%