Merck
  • Efficient delivery of signal-responsive gene carriers for disease-specific gene expression via bubble liposomes and sonoporation.
Efficient delivery of signal-responsive gene carriers for disease-specific gene expression via bubble liposomes and sonoporation.
Colloids and surfaces. B, Biointerfaces (2017-09-17)
Akira Tsuchiya, Jeong-Hun Kang, Takeshi Mori, Yuki Naritomi, Satoshi Kushio, Takuro Niidome, Katsuro Tachibana, Yoko Takahashi, Yoichi Negishi, Yusuke Oda, Ryo Suzuki, Kazuo Maruyama, Yoshiki Katayama
ZUSAMMENFASSUNG
Sonoporation is a promising method to intracellularly deliver synthetic gene carriers that have lower endocytotic uptake than viral carriers. Here, we applied sonoporation to deliver genes via polyethylene glycol (PEG)-grafted polymeric carriers that specifically respond to hyperactivated protein kinase A (PKA). PEG-grafted polymeric carrier/DNA polyplexes were not efficiently delivered into cells via the endocytotic pathway because of the hydrophilic PEG layer surrounding the polyplexes. However, the delivery of polyplexes into cells was significantly increased by sonoporation. The delivered polyplexes exhibited PKA-responsive transgene expression in PKA-overexpressing cells, but not in cells with low PKA activation. These results show that the sonoporation-mediated delivery of PEG-modified PKA-responsive polyplexes is a promising approach for safely applying gene therapy to abnormal cells with hyperactivated PKA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triisopropylsilan, 98%
Sigma-Aldrich
Acrylsäure N-Hydroxysuccinimid-Ester, ≥90% (HPLC)