Merck
Differentiation of neural stem cells into Schwann-like cells in vitro.
Biochemical and biophysical research communications (2010-10-06)
Lei Tong, Lili Ji, Zhenyu Wang, Xiaojie Tong, Lixin Zhang, Xiaohong Sun
ZUSAMMENFASSUNG
Neural stem cells (NSCs) are multipotent stem cells that have the potential to differentiate into different cells of the neural lineage like neurons, astrocytes and oligodendrocytes. In the current work, we explored whether NSCs could be differentiated into functional Schwann-like cells, which has not been investigated up to date. NSCs were harvested from the hippocampus of rats and identified by single cell cloning and immunofluorescence staining. Then NSCs were treated with DMEM/F12 supplemented with forskolin, heregulin, bFGF, PDGF-AA and retinoic acid (RA). Differentiated NSCs (dNSCs) exhibited a spindle-like morphology similar to Schwann cells and expressed the glial markers p75 and S100. We also found that dNSCs could enhance neurite outgrowth when co-cultured with NG108-15 cells. These results indicated that NSCs, derived from hippocampus of rats, could differentiate into Schwann-like cells with morphological, phenotypic and functional similarities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium/Nährstoffmischung F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture