Merck
  • Alpha-Linolenic Acid from Perilla frutescens var. japonica Oil Protects Aβ-Induced Cognitive Impairment through Regulation of APP Processing and Aβ Degradation.
Alpha-Linolenic Acid from Perilla frutescens var. japonica Oil Protects Aβ-Induced Cognitive Impairment through Regulation of APP Processing and Aβ Degradation.
Journal of agricultural and food chemistry (2017-11-03)
Ah Young Lee, Myoung Hee Lee, Sanghyun Lee, Eun Ju Cho
ZUSAMMENFASSUNG
Alzheimer's disease (AD) is characterized by progressive cognitive and memory impairment. The major pathological hallmark of AD is the accumulation of amyloid beta (Aβ), which is produced from the amyloid precursor protein (APP) through cleavage of β- and γ-secretase. Recently, dietary plant oil containing ω-3 polyunsaturated fatty acid has become an attractive alternative source to fish oil containing eicosapentaenoic acid or docosahexaenoic acid (DHA). We investigated whether ALA isolated from perilla oil has direct effects on improvement of cognitive ability and molecular mechanisms in APP processing in comparison with DHA. In the present study, ICR mice were treated orally with ALA or DHA (100 mg/kg/day) for 14 days after i.c.v. injection of Aβ
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Ethyl-2-pyrrolidon, 98%