Merck
  • Anti-fibrotic effects of Salvia miltiorrhiza and Ligustrazine Injection on LX-2 cells involved with increased N-myc downstream-regulated gene 2 expression.
Anti-fibrotic effects of Salvia miltiorrhiza and Ligustrazine Injection on LX-2 cells involved with increased N-myc downstream-regulated gene 2 expression.
Chinese journal of integrative medicine (2016-12-10)
Jin Zheng, Li-Tian Ma, Qin-You Ren, Yue Hu, Yang Bai, Huan Bian, Yi Zhang, Yong-Chun Zhou, Ming-Hui Yang
ZUSAMMENFASSUNG
To investigate the effects of Salvia miltiorrhiza and Ligustrazine Injection (SML) on proliferation and apoptosis of human hepatic stellate cell LX-2 and the expression of N-myc downstreamregulated gene 2 (NDRG2, a tumor suppressor gene). HSCs from the LX-2 cell line were cultured in vitro. The proliferative state of different initial LX-2 cell numbers was measured using a 3-(4,5-dimethylthiazol- 2-yl)-2,5-diphenyltetrazolium bromide (MTT) colorimetric assay. LX-2 cells were plated in 96-well plates at an approximate density of 2.50×10 With the exception of the 1 and 2 μL/mL concentrations, 4 and 8 μL/mL SML inhibited cell proliferation in a concentration-dependent manner at 24 and 48 h (P<0.05). With the exception of the 1 and 2 μL/mL concentrations, the NDRG2 expression level was greatly increased in a concentration-dependent manner. However, the level of β-catenin was unaffected. SML inhibit LX-2 cell proliferation in a concentration-dependent manner, and the mechanism may be associated with NDRG2 over-expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O