Merck
UiO-66-NH
ACS applied materials & interfaces (2017-11-23)
Dennis T Lee, Junjie Zhao, Christopher J Oldham, Gregory W Peterson, Gregory N Parsons
ZUSAMMENFASSUNG
Metal-organic frameworks (MOFs) chemically bound to polymeric microfibrous textiles show promising performance for many future applications. In particular, Zr-based UiO-66-family MOF-textiles have been shown to catalytically degrade highly toxic chemical warfare agents (CWAs), where favorable MOF/polymer bonding and adhesion are attained by placing a nanoscale metal-oxide layer on the polymer fiber preceding MOF growth. To date, however, the nucleation mechanism of Zr-based MOFs on different metal oxides and how product performance is affected are not well understood. Herein, we provide new insight into how different inorganic nucleation films (i.e., Al
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminoterephthalsäure, 99%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)