- Photodegradative fate and potential phototoxic products of bromocarbazoles and chlorocarbazoles in water.
Photodegradative fate and potential phototoxic products of bromocarbazoles and chlorocarbazoles in water.
Environmental science and pollution research international (2017-10-06)
John Mumbo, Dominik Deyerling, Bernhard Henkelmann, Gerd Pfister, Karl-Werner Schramm
PMID28980215
ZUSAMMENFASSUNG
Bromocarbazoles and chlorocarbazoles are emerging environmental contaminants that have been reported to be persistent and possessing dioxin-like toxicity; however, their photodegradative fate in water is unknown. The photodegradation of 3-bromocarbazole, 3-chlorocarbazole, and 3,6-dichlorocarbazole was determined in ultrapure water. They proceeded by direct photolysis and followed first-order kinetics. The rate constants (k) were 0.4838, 0.3454, and 0.4422 h