Merck
In vivo proton observed carbon edited (POCE)
Magnetic resonance in medicine (2017-05-13)
Chathura Kumaragamage, Dan Madularu, Axel P Mathieu, Henk De Feyter, M Natasha Rajah, Jamie Near
ZUSAMMENFASSUNG
In vivo carbon-13 ( A The coil provided excellent localization, uniform spin nutation, and sensitivity. A convenient and sensitive platform to study energy metabolism and neurotransmitter cycling is presented. Magn Reson Med 79:628-635, 2018. © 2017 International Society for Magnetic Resonance in Medicine.