Merck
  • Spatial structure of TLR4 transmembrane domain in bicelles provides the insight into the receptor activation mechanism.
Spatial structure of TLR4 transmembrane domain in bicelles provides the insight into the receptor activation mechanism.
Scientific reports (2017-08-02)
Konstantin S Mineev, Sergey A Goncharuk, Marina V Goncharuk, Pavel E Volynsky, Ekaterina V Novikova, Alexander S Aresinev
ZUSAMMENFASSUNG
Toll-like receptors (TLRs) play a key role in the innate and adaptive immune systems. While a lot of structural data is available for the extracellular and cytoplasmic domains of TLRs, and a model of the dimeric full-length TLR3 receptor in the active state was build, the conformation of the transmembrane (TM) domain and juxtamembrane regions in TLR dimers is still unclear. In the present work, we study the transmembrane and juxtamembrane parts of human TLR4 receptor using solution NMR spectroscopy in a variety of membrane mimetics, including phospholipid bicelles. We show that the juxtamembrane hydrophobic region of TLR4 includes a part of long TM α-helix. We report the dimerization interface of the TM domain and claim that long TM domains with transmembrane charged aminoacids is a common feature of human toll-like receptors. This fact is analyzed from the viewpoint of protein activation mechanism, and a model of full-length TLR4 receptor in the dimeric state has been proposed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumacetat -Lösung, BioUltra, Molecular Biology, ~1 M in H2O