Merck
  • Subsequent bone and metabolic responses of broilers to high-non-phytate phosphorus diets in the starter period.
Subsequent bone and metabolic responses of broilers to high-non-phytate phosphorus diets in the starter period.
British poultry science (2017-05-10)
N Baradaran, M H Shahir, Z Asadi Kermani
ZUSAMMENFASSUNG
1. An experiment was performed to elucidate the subsequent effects of high-non-phytate phosphorus (NPP) diets on growth performance, blood metabolites, bone characteristics and P retention of broilers fed on low-NPP grower diets. The 42-d study was designed as a 2 × 2 × 2 + 1 factorial, which included two starter NPP concentrations (4.5 and 5.5 g/kg; d 0-21), two grower NPP concentrations (1.5 and 2.3 g/kg; d 22-42), with or without phytase (1000 FTU/kg), with a reference diet containing an adequate NPP concentration over the course of the trial. 2. In the starter period, growth performance and P retention were not affected by experimental diets. The high-NPP diet increased plasma P concentration, increased tibia ash and tibia P contents and decreased plasma alkaline phosphatase (ALP) activity at d 21. 3. No significant interaction was observed between NPP concentrations in the starter and grower periods and phytase. The main effect data indicated that the increase in NPP concentration in the starter diets had no effects on growth performance in the grower period and overall. The high-NPP diet in the early stage of growth reduced plasma P concentration, plasma ALP activity and tibia ash content at d 42. The main effect data also showed that exogenous phytase increased body weight gain in the grower period and overall. 4. It can be concluded that feeding increased NPP diets have no effects on growth performance in the starter period. This feeding strategy results in negative effects on plasma P concentration and bone ash content at d 42. Also, exogenous phytase is effective in improving growth performance, bone characteristics and apparent P retention of growing broilers fed diets that are inadequate in phosphorus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)