Merck
Nfu1 Mediated ROS Removal Caused by Cd Stress in
Frontiers in physiology (2018-01-13)
Guang Qian, Yongbo Bao, Chenghua Li, Qingqing Xie, Meng Lu, Zhihua Lin
ZUSAMMENFASSUNG
The blood clam
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human NFU1