- Enzyme-Mimetic Antioxidant Luminescent Nanoparticles for Highly Sensitive Hydrogen Peroxide Biosensing.
Enzyme-Mimetic Antioxidant Luminescent Nanoparticles for Highly Sensitive Hydrogen Peroxide Biosensing.
ACS nano (2017-11-29)
Anna Pratsinis, Georgios A Kelesidis, Stefanie Zuercher, Frank Krumeich, Sreenath Bolisetty, Raffaele Mezzenga, Jean-Christophe Leroux, Georgios A Sotiriou
PMID29182310
ZUSAMMENFASSUNG
Hydrogen peroxide (H