Merck
GTRAP3-18 regulates food intake and body weight by interacting with pro-opiomelanocortin.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-09-15)
Koji Aoyama, Wattanaporn Bhadhprasit, Masahiko Watabe, Fan Wang, Nobuko Matsumura, Toshio Nakaki
ZUSAMMENFASSUNG
Pro-opiomelanocortin (POMC)-expressing neurons provide α-melanocyte-stimulating hormone (α-MSH), which stimulates melanocortin 4 receptor to induce hypophagia by AMPK inhibition in the hypothalamus. α-MSH is produced by POMC cleavage in secretory granules and released. However, it is not known yet whether any posttranscriptional regulatory mechanism of POMC signaling exists upstream of the secretory granules in neurons. Here we show that glutamate transporter-associated protein 3-18 (GTRAP3-18), an anchor protein that retains interacting proteins in the endoplasmic reticulum, is a critical regulator of food intake and body weight by interacting with POMC.