Merck
  • Eudesmane Sesquiterpenes from Verbesina lanata with Inhibitory Activity against Grapevine Downy Mildew.
Eudesmane Sesquiterpenes from Verbesina lanata with Inhibitory Activity against Grapevine Downy Mildew.
Journal of natural products (2017-12-05)
Justine Ramseyer, Barbara Thuerig, Maria De Mieri, Hans-Jakob Schärer, Thomas Oberhänsli, Mahabir P Gupta, Lucius Tamm, Matthias Hamburger, Olivier Potterat
ZUSAMMENFASSUNG
An in-house library of more than 3000 extracts of plant and fungal origin was screened against some major plant pathogens. As one of the hits, an ethyl acetate extract from inflorescences of Verbesina lanata showed significant inhibitory activity in vitro against grapevine downy mildew (Plasmopara viticola), with a MIC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-hydroxid, technical grade