Merck
  • Protein extraction into the bicontinuous microemulsion phase of a Water/SDS/pentanol/dodecane winsor-III system: Effect on nanostructure and protein conformation.
Protein extraction into the bicontinuous microemulsion phase of a Water/SDS/pentanol/dodecane winsor-III system: Effect on nanostructure and protein conformation.
Colloids and surfaces. B, Biointerfaces (2017-09-19)
Douglas G Hayes, Ran Ye, Rachel N Dunlap, Matthew J Cuneo, Sai Venkatesh Pingali, Hugh M O'Neill, Volker S Urban
ZUSAMMENFASSUNG
Bicontinuous microemulsions (BμEs), consisting of water and oil nanodomains separated by surfactant monolayers of near-zero curvature, are potentially valuable systems for purification and delivery of biomolecules, for hosting multiphasic biochemical reactions, and as templating media for preparing nanomaterials. We formed Winsor-III systems by mixing aqueous protein and sodium dodecyl sulfate (SDS) solutions with dodecane and 1-pentanol (cosurfactant) to efficiently extract proteins into the middle (BμE) phase. Bovine serum albumin (BSA) and cytochrome c partitioned to the BμE phase at 64% and 81% efficiency, respectively, producing highly concentrated protein solutions (32 and 44gL