- Inhibition of cancer cell migration with CuS@ mSiO
Inhibition of cancer cell migration with CuS@ mSiO
International journal of nanomedicine (2018-01-11)
Guoying Deng, Feng Zhou, Zizheng Wu, Fei Zhang, Kerun Niu, Yingjie Kang, Xijian Liu, Qiugen Wang, Yin Wang, Qian Wang
PMID29317819
ZUSAMMENFASSUNG
The metastasis of cancer cells is a vital aspect of disease progression and therapy. Although a few nanoparticles (NPs) aimed at controlling metastasis in cancer therapy have been reported, the NPs are normally combined with drugs, yet the direct therapeutic effects of the NPs are not reported. To study the direct influence of NPs on cancer metastasis, the potential suppression capacity of CuS@mSiO
MATERIALIEN