Merck
Development of a Terpenoid-Production Platform in Streptomyces reveromyceticus SN-593.
ACS synthetic biology (2017-10-12)
Ammara Khalid, Hiroshi Takagi, Suresh Panthee, Makoto Muroi, Joe Chappell, Hiroyuki Osada, Shunji Takahashi
ZUSAMMENFASSUNG
Terpenoids represent the largest class of natural products, some of which are resources for pharmaceuticals, fragrances, and fuels. Generally, mass production of valuable terpenoid compounds is hampered by their low production levels in organisms and difficulty of chemical synthesis. Therefore, the development of microbial biosynthetic platforms represents an alternative approach. Although microbial terpenoid-production platforms have been established in Escherichia coli and yeast, an optimal platform has not been developed for Streptomyces species, despite the large capacity to produce secondary metabolites, such as polyketide compounds. To explore this potential, we constructed a terpenoid-biosynthetic platform in Streptomyces reveromyceticus SN-593. This strain is unique in that it harbors the mevalonate gene cluster enabling the production of furaquinocin, which can be controlled by the pathway specific regulator Fur22. We simultaneously expressed the mevalonate gene cluster and subsequent terpenoid-biosynthetic genes under the control of Fur22. To achieve improved fur22 gene expression, we screened promoters from S. reveromyceticus SN-593. Our results showed that the promoter associated with rvr2030 gene enabled production of 212 ± 20 mg/L botryococcene to levels comparable to those previously reported for other microbial hosts. Given that the rvr2030 gene encodes for an enzyme involved in the primary metabolism, these results suggest that optimized expression of terpenoid-biosynthetic genes with primary and secondary metabolism might be as important for high yields of terpenoid compounds as is the absolute expression level of a target gene(s).