With the aim to obtain an injectable bioactive scaffold that can accelerate bone formation in sinus lift augmentation, in bony void and fracture repair, we have developed a three-dimensional (3D) jelly collagen containing lysophosphatidic acid (LPA) and 1α,25-dihydroxyvitamin D3 (1,25D3). Using an in vitro 3D culture model of bone fracture, we show that the contraction of the collagen gel is mediated by Rho-kinase activation in osteoblasts. The gel contraction showed dependence on cell concentration and was increased by LPA, which favored apposition and fastening of bone fragments approach. LPA was shown to act through actin cytoskeleton reorganization and myosin light chain phosphorylation of human primary osteoblasts (hOB). Moreover, LPA conferred osteoconductive properties as evidenced by the induction of proliferation, differentiation, and migration of hOB. The addition of 1,25D3 did not enhance cell-mediated gel contraction, but stimulated the maturation of hOB in vitro through the production of extracellular matrix of higher quality. On the basis of these observations, the collagen gel enriched with LPA and 1,25D3 described herein can be considered an injectable natural scaffold that allows the migration of cells from the side of bone defect and a promising candidate to accelerate bone growth and fracture healing.
Tissue engineering. Part A (2017-05-23)
Michela Bosetti, Alessia Borrone, Massimiliano Leigheb, V Prasad Shastri, Mario Cannas
PMID28530130
ZUSAMMENFASSUNG
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.