Merck
A Method for the Acute and Rapid Degradation of Endogenous Proteins.
Cell (2017-11-21)
Dean Clift, William A McEwan, Larisa I Labzin, Vera Konieczny, Binyam Mogessie, Leo C James, Melina Schuh
ZUSAMMENFASSUNG
Methods for the targeted disruption of protein function have revolutionized science and greatly expedited the systematic characterization of genes. Two main approaches are currently used to disrupt protein function: DNA knockout and RNA interference, which act at the genome and mRNA level, respectively. A method that directly alters endogenous protein levels is currently not available. Here, we present Trim-Away, a technique to degrade endogenous proteins acutely in mammalian cells without prior modification of the genome or mRNA. Trim-Away harnesses the cellular protein degradation machinery to remove unmodified native proteins within minutes of application. This rapidity minimizes the risk that phenotypes are compensated and that secondary, non-specific defects accumulate over time. Because Trim-Away utilizes antibodies, it can be applied to a wide range of target proteins using off-the-shelf reagents. Trim-Away allows the study of protein function in diverse cell types, including non-dividing primary cells where genome- and RNA-targeting methods are limited.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N6,2′-O-Dibutyryladenosin 3′,5′-zyklisches Monophosphat Natriumsalz Monohydrat, ≥96% (HPLC), powder
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-γ-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone GTU-88, ascites fluid
Sigma-Aldrich
Normale Kaninchen-IgG-Antikörper, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Normales Maus-IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Supelco
Zertifizierte sek. Standard-Referenz-Pufferlösung, potassium hydrogen phthalate, directly traceable to primary SRM from NIST/PTB pH (S) = 4.005 (25°C) Certipur®
Sigma-Aldrich
Anti-PCM1-Antikörper in Kaninchen hergestellte Antikörper, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-Polyglutamines antibody, Mouse monoclonal, ~2 mg/mL, clone 3B5H10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Kaninchen-Leichtkette der Maus, HRP-Konjugat-Antikörper, Chemicon®, from mouse
Sigma-Aldrich
Anti-Maus-Leichtketten-Antikörper der Ziege, HRP-Konjugat, Chemicon®, from goat
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Raptor-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-KIF11 antibody produced in rabbit, affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution