Merck
  • Multiple signaling kinases target Mrc1 to prevent genomic instability triggered by transcription-replication conflicts.
Multiple signaling kinases target Mrc1 to prevent genomic instability triggered by transcription-replication conflicts.
Nature communications (2018-01-27)
Alba Duch, Berta Canal, Sonia I Barroso, María García-Rubio, Gerhard Seisenbacher, Andrés Aguilera, Eulàlia de Nadal, Francesc Posas
ZUSAMMENFASSUNG
Conflicts between replication and transcription machineries represent a major source of genomic instability and cells have evolved strategies to prevent such conflicts. However, little is known regarding how cells cope with sudden increases of transcription while replicating. Here, we report the existence of a general mechanism for the protection of genomic integrity upon transcriptional outbursts in S phase that is mediated by Mrc1. The N-terminal phosphorylation of Mrc1 blocked replication and prevented transcription-associated recombination (TAR) and genomic instability during stress-induced gene expression in S phase. An unbiased kinome screening identified several kinases that phosphorylate Mrc1 at the N terminus upon different environmental stresses. Mrc1 function was not restricted to environmental cues but was also required when unscheduled transcription was triggered by low fitness states such as genomic instability or slow growth. Our data indicate that Mrc1 integrates multiple signals, thereby defining a general safeguard mechanism to protect genomic integrity upon transcriptional outbursts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution