Merck
  • Epigenetically induced ectopic expression of UNCX impairs the proliferation and differentiation of myeloid cells.
Epigenetically induced ectopic expression of UNCX impairs the proliferation and differentiation of myeloid cells.
Haematologica (2017-04-16)
Giulia Daniele, Giorgia Simonetti, Caterina Fusilli, Ilaria Iacobucci, Angelo Lonoce, Antonio Palazzo, Mariana Lomiento, Fabiana Mammoli, Renè Massimiliano Marsano, Elena Marasco, Vilma Mantovani, Hilmar Quentmeier, Hans G Drexler, Jie Ding, Orazio Palumbo, Massimo Carella, Niroshan Nadarajah, Margherita Perricone, Emanuela Ottaviani, Carmen Baldazzi, Nicoletta Testoni, Cristina Papayannidis, Sergio Ferrari, Tommaso Mazza, Giovanni Martinelli, Clelia Tiziana Storlazzi
ZUSAMMENFASSUNG
We here describe a leukemogenic role of the homeobox gene