Merck
  • The small G protein Arf6 expressed in keratinocytes by HGF stimulation is a regulator for skin wound healing.
The small G protein Arf6 expressed in keratinocytes by HGF stimulation is a regulator for skin wound healing.
Scientific reports (2017-04-22)
Yuki Miura, Van Ngo Thai Bich, Momoko Furuya, Hiroshi Hasegawa, Satoru Takahashi, Naohiro Katagiri, Tsunaki Hongu, Yuji Funakoshi, Norihiko Ohbayashi, Yasunori Kanaho
ZUSAMMENFASSUNG
The earlier step of cutaneous wound healing process, re-epithelialization of the wounded skin, is triggered by a variety of growth factors. However, molecular mechanisms through which growth factors trigger skin wound healing are less understood. Here, we demonstrate that hepatocyte growth factor (HGF)/c-Met signaling-induced expression of the small G protein Arf6 mRNA in keratinocytes is essential for the skin wound healing. Arf6 mRNA expression was dramatically induced in keratinocytes at the wounded skin, which was specifically suppressed by the c-Met inhibitor. Wound healing of the skin was significantly delayed in keratinocyte-specific Arf6 conditional knockout mice. Furthermore, Arf6 deletion from keratinocytes remarkably suppressed HGF-stimulated cell migration and peripheral membrane ruffle formation, but did not affect skin morphology and proliferation/differentiation of keratinocytes. These results are consistent with the notion that Arf6 expressed in skin keratinocytes through the HGF/c-Met signaling pathway in response to skin wounding plays an important role in skin wound healing by regulating membrane dynamics-based motogenic cellular function of keratinocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonaler Anti-BrdU in Maus hergestellte Antikörper, clone BU-33, ascites fluid, Immunohistology Grade