Merck
  • Variants in the Oxidoreductase PYROXD1 Cause Early-Onset Myopathy with Internalized Nuclei and Myofibrillar Disorganization.
Variants in the Oxidoreductase PYROXD1 Cause Early-Onset Myopathy with Internalized Nuclei and Myofibrillar Disorganization.
American journal of human genetics (2016-10-18)
Gina L O'Grady, Heather A Best, Tamar E Sztal, Vanessa Schartner, Myriam Sanjuan-Vazquez, Sandra Donkervoort, Osorio Abath Neto, Roger Bryan Sutton, Biljana Ilkovski, Norma Beatriz Romero, Tanya Stojkovic, Jahannaz Dastgir, Leigh B Waddell, Anne Boland, Ying Hu, Caitlin Williams, Avnika A Ruparelia, Thierry Maisonobe, Anthony J Peduto, Stephen W Reddel, Monkol Lek, Taru Tukiainen, Beryl B Cummings, Himanshu Joshi, Juliette Nectoux, Susan Brammah, Jean-François Deleuze, Viola Oorschot Ing, Georg Ramm, Didem Ardicli, Kristen J Nowak, Beril Talim, Haluk Topaloglu, Nigel G Laing, Kathryn N North, Daniel G MacArthur, Sylvie Friant, Nigel F Clarke, Robert J Bryson-Richardson, Carsten G Bönnemann, Jocelyn Laporte, Sandra T Cooper
ZUSAMMENFASSUNG
This study establishes PYROXD1 variants as a cause of early-onset myopathy and uses biospecimens and cell lines, yeast, and zebrafish models to elucidate the fundamental role of PYROXD1 in skeletal muscle. Exome sequencing identified recessive variants in PYROXD1 in nine probands from five families. Affected individuals presented in infancy or childhood with slowly progressive proximal and distal weakness, facial weakness, nasal speech, swallowing difficulties, and normal to moderately elevated creatine kinase. Distinctive histopathology showed abundant internalized nuclei, myofibrillar disorganization, desmin-positive inclusions, and thickened Z-bands. PYROXD1 is a nuclear-cytoplasmic pyridine nucleotide-disulphide reductase (PNDR). PNDRs are flavoproteins (FAD-binding) and catalyze pyridine-nucleotide-dependent (NAD/NADH) reduction of thiol residues in other proteins. Complementation experiments in yeast lacking glutathione reductase glr1 show that human PYROXD1 has reductase activity that is strongly impaired by the disease-associated missense mutations. Immunolocalization studies in human muscle and zebrafish myofibers demonstrate that PYROXD1 localizes to the nucleus and to striated sarcomeric compartments. Zebrafish with ryroxD1 knock-down recapitulate features of PYROXD1 myopathy with sarcomeric disorganization, myofibrillar aggregates, and marked swimming defect. We characterize variants in the oxidoreductase PYROXD1 as a cause of early-onset myopathy with distinctive histopathology and introduce altered redox regulation as a primary cause of congenital muscle disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-α-Aktinin (Sarkomer) in Maus hergestellte Antikörper, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus, clone B-5-1-2, purified from hybridoma cell culture