Merck
Differential Responses of Human Fetal Brain Neural Stem Cells to Zika Virus Infection.
Stem cell reports (2017-02-22)
Erica L McGrath, Shannan L Rossi, Junling Gao, Steven G Widen, Auston C Grant, Tiffany J Dunn, Sasha R Azar, Christopher M Roundy, Ying Xiong, Deborah J Prusak, Bradford D Loucas, Thomas G Wood, Yongjia Yu, Ildefonso Fernández-Salas, Scott C Weaver, Nikos Vasilakis, Ping Wu
ZUSAMMENFASSUNG
Zika virus (ZIKV) infection causes microcephaly in a subset of infants born to infected pregnant mothers. It is unknown whether human individual differences contribute to differential susceptibility of ZIKV-related neuropathology. Here, we use an Asian-lineage ZIKV strain, isolated from the 2015 Mexican outbreak (Mex1-7), to infect primary human neural stem cells (hNSCs) originally derived from three individual fetal brains. All three strains of hNSCs exhibited similar rates of Mex1-7 infection and reduced proliferation. However, Mex1-7 decreased neuronal differentiation in only two of the three stem cell strains. Correspondingly, ZIKA-mediated transcriptome alterations were similar in these two strains but significantly different from that of the third strain with no ZIKV-induced neuronal reduction. This study thus confirms that an Asian-lineage ZIKV strain infects primary hNSCs and demonstrates a cell-strain-dependent response of hNSCs to ZIKV infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Mikrotubuli-assoziiertes Protein 2 (MAP2)-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-(saures) Gliafilamentprotein in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution