Merck
Sucralose activates an ERK1/2-ribosomal protein S6 signaling axis.
FEBS open bio (2017-02-09)
Marcy L Guerra, Michael A Kalwat, Kathleen McGlynn, Melanie H Cobb
ZUSAMMENFASSUNG
The sweetener sucralose can signal through its GPCR receptor to induce insulin secretion from pancreatic β cells, but the downstream signaling pathways involved are not well-understood. Here we measure responses to sucralose, glucagon-like peptide 1, and amino acids in MIN6 β cells. Our data suggest a signaling axis, whereby sucralose induces calcium and cAMP, activation of ERK1/2, and site-specific phosphorylation of ribosomal protein S6. Interestingly, sucralose acted independently of mTORC1 or ribosomal S6 kinase (RSK). These results suggest that sweeteners like sucralose can influence β-cell responses to secretagogues like glucose through metabolic as well as GPCR-mediated pathways. Future investigation of novel sweet taste receptor signaling pathways in β cells will have implications for diabetes and other emergent fields involving these receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale Anti-MAP-Kinase, aktiviert (Diphosphorylierter ERK-1&2) in Maus hergestellte Antikörper, clone MAPK-YT, ascites fluid