- Thalidomide inhibits proliferation and epithelial-mesenchymal transition by modulating CD133 expression in pancreatic cancer cells.
Thalidomide inhibits proliferation and epithelial-mesenchymal transition by modulating CD133 expression in pancreatic cancer cells.
Oncology letters (2018-01-19)
Congying Chen, Ge Yu, Wenqin Xiao, Miao Xing, Jianbo Ni, Rong Wan, Guoyong Hu
PMID29344263
ZUSAMMENFASSUNG
Pancreatic cancer is a solid malignancy with a high mortality rate, on account of the high incidence of metastasis at the time of detection. The aggressiveness of pancreatic cancer may be partly driven by cancer stem cells (CSCs), which are characterized by the ability to self-renew and recapitulate tumors in the ectopic setting. However, although a number of drugs targeting CSCs are currently under clinical investigation, few effective drugs have been developed. The present study demonstrated that thalidomide inhibited cell proliferation and metastasis in pancreatic cancer cell lines through the inhibition of epithelial mesenchymal transition. The effect of thalidomide was more pronounced in cluster of differentiation 133 (CD133)