Merck
  • Roles of TGFβ and FGF signals during growth and differentiation of mouse lens epithelial cell in vitro.
Roles of TGFβ and FGF signals during growth and differentiation of mouse lens epithelial cell in vitro.
Scientific reports (2017-08-06)
Dong Wang, Eddie Wang, Kelsey Liu, Chun-Hong Xia, Song Li, Xiaohua Gong
ZUSAMMENFASSUNG
Transforming growth factor β (TGFβ) and fibroblast growth factor (FGF) signaling pathways play important roles in the proliferation and differentiation of lens epithelial cells (LECs) during development. Low dosage bFGF promotes cell proliferation while high dosage induces differentiation. TGFβ signaling regulates LEC proliferation and differentiation as well, but also promotes epithelial-mesenchymal transitions that lead to cataracts. Thus far, it has been difficult to recapitulate the features of germinative LECs in vitro. Here, we have established a LEC culture protocol that uses SB431542 (SB) compound to inhibit TGFβ/Smad activation, and found that SB treatment promoted mouse LEC proliferation, maintained LECs' morphology and distinct markers including N-cadherin, c-Maf, Prox1, and αA-, αB-, and β-crystallins. In contrast, low-dosage bFGF was unable to sustain those markers and, combined with SB, altered LECs' morphology and β-crystallin expression. We further found that Matrigel substrate coatings greatly increased cell proliferation and uniquely affected β-crystallin expression. Cultured LECs retained the ability to differentiate into γ-crystallin-positive lentoids by high-dosage bFGF treatment. Thus, a suppression of TGFβ/Smad signaling in vitro is critical to maintaining characteristic features of mouse LECs, especially expression of the key transcription factors c-Maf and Prox1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-Prox-1-Antikörper, serum, Chemicon®