- Proteomics profiling of interactome dynamics by colocalisation analysis (COLA).
Proteomics profiling of interactome dynamics by colocalisation analysis (COLA).
Molecular bioSystems (2016-11-09)
Faraz K Mardakheh, Heba Z Sailem, Sandra Kümper, Christopher J Tape, Ryan R McCully, Angela Paul, Sara Anjomani-Virmouni, Claus Jørgensen, George Poulogiannis, Christopher J Marshall, Chris Bakal
PMID27824369
ZUSAMMENFASSUNG
Localisation and protein function are intimately linked in eukaryotes, as proteins are localised to specific compartments where they come into proximity of other functionally relevant proteins. Significant co-localisation of two proteins can therefore be indicative of their functional association. We here present COLA, a proteomics based strategy coupled with a bioinformatics framework to detect protein-protein co-localisations on a global scale. COLA reveals functional interactions by matching proteins with significant similarity in their subcellular localisation signatures. The rapid nature of COLA allows mapping of interactome dynamics across different conditions or treatments with high precision.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture