Merck
Context-dependent autoprocessing of human immunodeficiency virus type 1 protease precursors.
PloS one (2018-01-18)
ChihFeng Tien, Liangqun Huang, Susan M Watanabe, Jordan T Speidel, Carol A Carter, Chaoping Chen
ZUSAMMENFASSUNG
HIV-1 protease autoprocessing is responsible for liberation of free mature protease (PR) from the Gag-Pol polyprotein precursor. A cell-based model system was previously developed to examine the autoprocessing mechanism of fusion precursors carrying the p6*-PR miniprecursor sandwiched between various proteins or epitopes. We here report that precursor autoprocessing is context-dependent as its activity and outcomes can be modulated by sequences upstream of p6*-PR. This was exemplified by the 26aa maltose binding protein (MBP) signal peptide (SigP) when placed at the N-terminus of a fusion precursor. The mature PRs released from SigP-carrying precursors are resistant to self-degradation whereas those released from SigP-lacking fusion precursors are prone to self-degradation. A H69D mutation in PR abolished autoprocessing of SigP-containing fusion precursors whereas it only partially suppressed autoprocessing of fusion precursors lacking SigP. An autoprocessing deficient GFP fusion precursor with SigP exhibited a subcellular distribution pattern distinct from the one without it in transfected HeLa cells. Furthermore, a SigP fusion precursor carrying a substitution at the P1 position released the mature PR and PR-containing fragments that were different from those released from the precursor carrying the same mutation but lacking SigP. We also examined autoprocessing outcomes in viral particles produced by a NL4-3 derived proviral construct and demonstrated the existence of several PR-containing fragments along with the mature PR. Some of these resembled the SigP precursor autoprocessing outcomes. This finding of context-dependent modulation reveals the complexity of precursor autoprocessing regulation that most likely accompanies sequence variation imposed by the evolution of the upstream Gag moiety.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-Glutathion-S-Transferase (GST) in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution