Merck
  • Stereoselective synthesis of Boc-protected cis and trans-4-trifluoromethylprolines by asymmetric hydrogenation reactions.
Stereoselective synthesis of Boc-protected cis and trans-4-trifluoromethylprolines by asymmetric hydrogenation reactions.
Angewandte Chemie (International ed. in English) (2002-05-03)
Juan R Del Valle, Murray Goodman
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(1,5-Cyclooctadien)(pyridin)(tricyclohexylphosphin)-iridium(I)-hexafluorphosphat, 85%
Sigma-Aldrich
(1,5-Cyclooctadien)(pyridin)(tricyclohexylphosphin)-iridium(I)-hexafluorphosphat, ≥99.0% (C)