- Kinetic modeling predicts a stimulatory role for ribosome collisions at elongation stall sites in bacteria.
Kinetic modeling predicts a stimulatory role for ribosome collisions at elongation stall sites in bacteria.
eLife (2017-05-13)
Michael A Ferrin, Arvind R Subramaniam
PMID28498106
ZUSAMMENFASSUNG
Ribosome stalling on mRNAs can decrease protein expression. To decipher ribosome kinetics at stall sites, we induced ribosome stalling at specific codons by starving the bacterium
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)