- Rapid and Efficient CRISPR/Cas9-Based Mating-Type Switching of
Rapid and Efficient CRISPR/Cas9-Based Mating-Type Switching of
G3 (Bethesda, Md.) (2017-11-19)
Ze-Xiong Xie, Leslie A Mitchell, Hui-Min Liu, Bing-Zhi Li, Duo Liu, Neta Agmon, Yi Wu, Xia Li, Xiao Zhou, Bo Li, Wen-Hai Xiao, Ming-Zhu Ding, Ying Wang, Ying-Jin Yuan, Jef D Boeke
PMID29150593
ZUSAMMENFASSUNG
Rapid and highly efficient mating-type switching of